Mesitylen ≥97.0%
Glass bottle, 25 mL
Katalog # TCIAT0470-25ML
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: 1,3,5-Trimetylbenzen
- CAS-nummer:108-67-8
- Formel:C₉H₁₂
- MDL-nummer:MFCD00008538
- Molecular Weight:120.19 g/mol
- Merck Index:14,05907
- Kokande Pt:164°C
- Smält Pt:–45 °C
- Densitet:0,8637 g/cm³ (20 °C)
- Flash Pt:50°C
- Förvaringstemperatur:Omgivande
- EINECS:203-604-4
- FN:2325
- ADR:3,III
Specifikationer
Specifikationstestresultat
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:25 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



