Paraformaldehyd ≥90.0%
Glass bottle, 500 g
Katalog # TCIAP0018-500G
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: Polyoxymethylene
- CAS-nummer:30525-89-4
- Formel:(CH₂O)n
- MDL-nummer:MFCD00133991
- Merck Index:14,07025
- Kokande Pt:135 - 136 °C (13 -rr)
- Smält Pt:123 °C
- Densitet:1,46 g/cm³ (20 °C)
- Flash Pt:70 °C
- Förvaringstemperatur:Omgivande
- FN:2213
- ADR:4.1,III
Specifikationer
Specifikationstestresultat
- Appearance:White powder to crystal
- Purity (Sodium Sulfite Method):min. 90.0 %
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



