1-Naphthaldehyde ≥95.0%
Glass bottle, 500 mL
TCIAN0002-500ML
: TCI
: α-Naphthaldehyde
- :66-77-3
- :C₁₁H₈O
- :MFCD00004003
- :156.18 g/mol
- :291°C
- :2 °C
- :1,14 g/cm³ (20 °C)
- :Omgivande
- :200-633-4
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
- Förpackningstyp:Glass bottle
- Förp.:500 mL



