Fluorbensen ≥99.0%
Glass bottle, 500 g
TCIAF0034-500G
: TCI
: Phenylfluoride
- :462-06-6
- :C₆H₅F
- :MFCD00000280
- :96.1 g/mol
- :14,04170
- :85°C
- :–41 °C
- :1,023 g/cm³ (20 °C)
- :–13°C
- :-13°C
- :Omgivande
- :207-321-7
- :2387
- :3,II
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
- Förpackningstyp:Glass bottle
- Förp.:500 g



