Fluorbensen ≥99.0%
Glass bottle, 25 g
Katalog # TCIAF0034-25G
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: Phenylfluoride
- CAS-nummer:462-06-6
- Formel:C₆H₅F
- MDL-nummer:MFCD00000280
- Molecular Weight:96.1 g/mol
- Merck Index:14,04170
- Kokande Pt:85°C
- Smält Pt:–41 °C
- Densitet:1,023 g/cm³ (20 °C)
- Flash Pt:–13°C
- Flash Pt:-13°C
- Förvaringstemperatur:Omgivande
- EINECS:207-321-7
- FN:2387
- ADR:3,II
Documents
Specifikationer
Specifikationstestresultat
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:25 g
- Cat. No.:TCIAF0034-25G
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



