Compensating Cables for Temperature Sensors
Compensating cable, type J, Svart/Vit, 10 m
WINKWFZJ0250
: WINKLER
Compensating cables in PTFE insulated, twisted leads.
- Färg:Svart/Vit
- Beskrivning:Compensating cable, type J
- Längd:10 m
- Seq:01
Compensating cables in PTFE insulated, twisted leads.