Compensating Cables for Temperature Sensors
Compensating cable, type K, Grön/vit, 10 m
Katalog # WINKWFZK0250
Leverantör: WINKLER
Gå till:
Product Details & Documents
Compensating cables in PTFE insulated, twisted leads.
Specifikationer
- Färg:Grön/vit
- Beskrivning:Compensating cable, type K
- Längd:10 m
- Seq:02