Heating tapes
Heating tape, 3,0 m length, 750 W
Katalog # WINKWBG00403000750
Leverantör: WINKLER
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:Heating tape, 3,0 m length, 750 W
- Seq:07