Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
BUFFER PACK PCR PHIRE REACTION 1 * 1 KIT
Katalog # F-524L
Leverantör:
FINNZYMES REAGENTS
Expandera bilden
BUFFER PACK PCR PHIRE REACTION 1 * 1 KIT
Katalog # F-524L
Leverantör:
FINNZYMES REAGENTS