Wire baskets, square
118×118×144 mm
Leverantör: DELTALAB
Gå till:
Product Details & Documents
Square baskets, made of stainless steel wire, with folding handles.
- Features a strong woven mesh base
- Good chemical resistance
- Good temperature resistance
- Comes without lid (to order baskets with lid please contact our customer service
Documents
Certificates
Sök efter LOT-nummerSpecifikationer
- Seq:01
- B×D×H:118×118×144 mm