Orders LinkContactUsLinkComponent
pH buffer solutions
pH buffer solutions
pH buffer standard solution with certificate of analysis, 125 ml, 4,01, 1
Katalog # HACHLZW9460.99
Leverantör:   Hach
pH buffer solutions
Katalog # HACHLZW9460.99
Leverantör:  Hach
CAS-nummer:  
pH buffer solutions
pH buffer solutions
pH buffer standard solution with certificate of analysis, 125 ml, 4,01, 1
CAS-nummer:  
Price
230,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Specifikationer
Specifikationer
  • Förp.:
    1
  • Beskrivning:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH-värde:
    4,01
Specifikationer