Tubes and systems for active sampling
Silica gel tubes, B/G
Katalog # DRAE8101861
Leverantör: Dräger
Gå till:
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
- High precision guaranteed
- Very reliable in use
- Improved colour depth and contrast using an optimised production method
Specifikationer
- Typ:B/G
- Beskrivning:Silica gel tubes
- Seq:21