Orders LinkContactUsLinkComponent
Test tubes
Test tubes
5 ml, 92 mm, 12 mm
Katalog # LPIT113140
Leverantör:   LP ITALIANA
Test tubes
Katalog # LPIT113140
Leverantör:  LP ITALIANA
Test tubes
Test tubes
5 ml, 92 mm, 12 mm
Price
3 600,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 3.600 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
PS, clear or PP, transparent. Universal tubes for a wide range of laboratory and production applications.
  • Good chemical resistance
  • Good temperature resistance
Ideal for sampling for laboratory testing.
Specifikationer
  • Ø utv.:
    12 mm
  • Kapacitet:
    5 ml
  • Längd:
    92 mm
Accessories
Produktinformation
  • Katalog #
    Material
    För ytterdiam.
    Tillgänglighet
    Price
  • Stoppers for RIA test tubes
    Stoppers for RIA test tubes
    LPIT112430
    PE
    11 mm
    Log in för att kolla tillgänglighet och ert pris
    1 * 12.000 Items - 3 875,00 kr