Counting chambers
Slides with four chambers, without grid, 3×3 mm
Katalog # 630-1538
Leverantör: Kova International
Gå till:
Product Details & Documents
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
- High optical quality through the composition acrylate
- Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
- Capillary action for reproducible precision
- Homogeneous suspension of sample sediments: Retained volume of 1 µl
- For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl
Specifikationer
- Typ:without grid
- B×D:3×3 mm
- Beskrivning:Slides with four chambers
- Cat. No.:630-1538