Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Accessories for centrifuges

Adapter for 15 ml tubes with screw cap, Ø 17×104 mm
Katalog # HETT1647
   Low Manufacturing Impact
Leverantör:   Hettich
Accessories for centrifuges
Katalog # HETT1647
Leverantör:  Hettich
   Low Manufacturing Impact
Accessories for centrifuges
Accessories for centrifuges

Adapter for 15 ml tubes with screw cap, Ø 17×104 mm

Price
3 050,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 6 Items
Product Details & Documents
Specifikationer
  • Beskrivning:
    Adapter for 15 ml tubes with screw cap, Ø 17×104 mm
  • Seq:
    62
  • Miljöcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Cat. No.:
    HETT1647