Orders LinkContactUsLinkComponent
Separating funnel, globe shaped
Separating funnel, globe shaped
250 ml, 29/32 NS
Katalog # KAVA632426351250
Leverantör:   Kavalier
Separating funnel, globe shaped
Katalog # KAVA632426351250
Leverantör:  Kavalier
Separating funnel, globe shaped
Separating funnel, globe shaped
250 ml, 29/32 NS
Price
4 725,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 2 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for separation of particular phases of the mixture. With standard ground joint stopper.
Specifikationer
  • NS-hylsa:
    29/32 NS
  • Kapacitet:
    250 ml