Order Entry
Sweden
Orders LinkContactUsLinkComponent
Centrifugrör

Centrifugrör

Tubes, with white screw cap, 15 ml, 16×100 mm, Med skruvlock, Vit, non sterile
Katalog # KART88303/03
Leverantör:   KARTELL
Centrifugrör
Katalog # KART88303/03
Leverantör:  KARTELL
Centrifugrör
Centrifugrör

Tubes, with white screw cap, 15 ml, 16×100 mm, Med skruvlock, Vit, non sterile

Price
3 575,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1.000 Items
Product Details & Documents

PP with quick closure PE screw cap.

  • Leak-proof
  • Can be centrifuged up to 4000×g
  • Cap is designed to avoid any contamination due to drops of liquid on the inner surface
Specifikationer
  • Stängningsstorlek:
    Med skruvlock
  • Ø×L:
    16×100 mm
  • Version:
    non sterile
  • Kapacitet:
    15 ml
  • Beskrivning:
    Tubes, with white screw cap
  • Seq:
    02
  • Lock, färg:
    Vit
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.