Gas sampling balloons
Gas sampling balloon, 230 mm, 5,0 L
Katalog # KART1234
Leverantör: KARTELL
Gå till:
Product Details & Documents
Gas collection bags made from latex.
Designed for general use in laboratories.
Beställningsinformation: Available in different volumes and diameters.
Documents
Specifikationer
- Ø:230 mm
- Volym:5,0 L
- Beskrivning:Gas sampling balloon