Filter vials, SINGLE StEP™
Nylon, 0,45 µm, Rosa
Katalog # 548-1410
Leverantör: THOMSON INSTRUMENT
Gå till:
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
- Compatible with both UHPLC and standard HPLC autosamplers
- Colour coded pre-slit caps for easy identification of membrane and porosity
- Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Specifikationer
- Membran:Nylon
- Porstorlek:0,45 µm
- Färg:Rosa
- Seq:10