Embedding cassettes
Embedding cassettes, Orange
Katalog # MARI5253109
Leverantör: MARIENFELD
Gå till:
Product Details & Documents
Plastic (POM).
- Resistant against the solvents usually used in laboratories
- Different colours offer the possibility of colour coding (for example for different users, priorities)
- One 45° inclined and two vertical marking areas
- Roughness of the marking areas optimized to allow labelling by different systems
Specifikationer
- Färg:Orange
- Beskrivning:Embedding cassettes
- Seq:02