Orders LinkContactUsLinkComponent
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
29/32 NS, GL 32, 13 - 15 mm
Katalog # LENZ01303229
Leverantör:   Lenz Laborglas
Adapters, glass, cone/screwthread
Katalog # LENZ01303229
Leverantör:  Lenz Laborglas
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
29/32 NS, GL 32, 13 - 15 mm
Price
805,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Documents
Specifikationer
  • Kärna:
    29/32 NS
  • Gänga:
    GL 32
  • Rör, ytterdiam.:
    13 - 15 mm
  • Seq:
    11