Orders LinkContactUsLinkComponent
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
19/26 NS, GL 25, 9 - 11 mm
Katalog # LENZ01302519
Leverantör:   Lenz Laborglas
Adapters, glass, cone/screwthread
Katalog # LENZ01302519
Leverantör:  Lenz Laborglas
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
19/26 NS, GL 25, 9 - 11 mm
Price
470,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Documents
Specifikationer
  • Kärna:
    19/26 NS
  • Gänga:
    GL 25
  • Rör, ytterdiam.:
    9 - 11 mm
  • Seq:
    08