Cell counter vials
Cell counter vial 1 with lid, for Coulter® and Hycel® cell counter systems, 25,00 ml, PS, Klar, Ja, 50 vials/tray, 20 trays
Katalog # RATI5515010
Leverantör: RATIOLAB
Gå till:
Produktdetaljer och dokument
Cell counter vials made of clear PS.
- High-precision design for use with robotic analytical systems
Specifikationer
- Med lock:Ja
- Förp.:50 vials/tray, 20 trays
- Kapacitet:25,00 ml
- Färg:Klar
- Beskrivning:Cell counter vial 1 with lid, for Coulter® and Hycel® cell counter systems
- Material:PS
- Seq:01