Order Entry
Sweden
Orders LinkContactUsLinkComponent
qPCR master mixes, Fast Plus EvaGreen®

qPCR master mixes, Fast Plus EvaGreen®

Fast Plus EvaGreen® qPCR master mix , 5×1 ml, 500, 5 mL
Katalog # 31020-1
   Safer Human and Environmental Health
Leverantör:   Biotium
qPCR master mixes, Fast Plus EvaGreen®
Katalog # 31020-1
Leverantör:  Biotium
   Safer Human and Environmental Health
CAS-nummer:  
qPCR master mixes, Fast Plus EvaGreen®
qPCR master mixes, Fast Plus EvaGreen®

Fast Plus EvaGreen® qPCR master mix , 5×1 ml, 500, 5 mL

CAS-nummer:  
Price
7 900,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 5 mL
Product Details & Documents

A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.

  • 2X qPCR master mix for fast cycling protocols
  • EvaGreen® dye-based
  • Contains Cheetah™ Taq hotstart DNA polymerase

Fast Plus EvaGreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.

Fast Plus Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.

Specifikationer

Specifikationer

  • Förp.:
    5 mL
  • Beskrivning:
    Fast Plus EvaGreen® qPCR master mix , 5×1 ml
  • No. of reactions:
    500
  • Cat. No.:
    31020-1
  • Applikation:
    qPCR dye
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.