Orders LinkContactUsLinkComponent
IgG Biotinylation kits, HOOK™
IgG Biotinylation kits, HOOK™
HOOK™ IgG Biotinylation Kit (Sulfhydryl), 8 reactions, 1 KIT
Katalog # 786-729
Leverantör:   G-Biosciences
IgG Biotinylation kits, HOOK™
Katalog # 786-729
Leverantör:  G-Biosciences
CAS-nummer:  
IgG Biotinylation kits, HOOK™
IgG Biotinylation kits, HOOK™
HOOK™ IgG Biotinylation Kit (Sulfhydryl), 8 reactions, 1 KIT
CAS-nummer:  
Price
6 415,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 KIT
Produktdetaljer och dokument
HOOK™ IgG biotinylation kits are designed for the biotinylation of IgG molecules by immobilising the IgG molecules on a solid support. Nickel chelating resin allows for the rapid removal of uncoupled biotin and therefore eliminates the need for further dialysis or desalting of the biotinylated antibody.
  • Simpler antibody biotinylation
  • Solid support technology eliminates dialysis/desalting
  • Suitable for 1 to 10mg antibody
  • PEG Biotin reagent for reduced steric hindrance and increased labelled antibody solubility
Specifikationer
Specifikationer
  • Förp.:
    1 KIT
  • Beskrivning:
    HOOK™ IgG Biotinylation Kit (Sulfhydryl), 8 reactions
Specifikationer