Orders LinkContactUsLinkComponent
IgG Biotinylation kits, HOOK™
HOOK™ IgG Biotinylation Kit (Amine), 8 reactions, 1 KIT
Catalog # 786-728
Supplier:   G-Biosciences
IgG Biotinylation kits, HOOK™
Catalog # 786-728
Supplier:  G-Biosciences
CAS Number:  
IgG Biotinylation kits, HOOK™
IgG Biotinylation kits, HOOK™
HOOK™ IgG Biotinylation Kit (Amine), 8 reactions, 1 KIT
CAS Number:  
Price
5 245,00 kr
Log in for availability
1 * 1 KIT
Product Details & Documents
HOOK™ IgG biotinylation kits are designed for the biotinylation of IgG molecules by immobilising the IgG molecules on a solid support. Nickel chelating resin allows for the rapid removal of uncoupled biotin and therefore eliminates the need for further dialysis or desalting of the biotinylated antibody.
  • Simpler antibody biotinylation
  • Solid support technology eliminates dialysis/desalting
  • Suitable for 1 to 10mg antibody
  • PEG Biotin reagent for reduced steric hindrance and increased labelled antibody solubility
Specifications
Specifications
  • Förp.:
    1 KIT
  • Beskrivning:
    HOOK™ IgG Biotinylation Kit (Amine), 8 reactions
Specifications