qPCR master mixes, Fast Plus EvaGreen®
Fast Plus EvaGreen® qPCR master mix with low Rox, 1 ml, 100, 1 mL
Katalog # 31014-T
Leverantör: Biotium
qPCR master mixes, Fast Plus EvaGreen®
Katalog # 31014-T
Leverantör: Biotium
CAS-nummer:
Gå till:
Product Details & Documents
A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.
- 2X qPCR master mix for fast cycling protocols
- EvaGreen® dye-based
- Contains Cheetah™ Taq hotstart DNA polymerase
Fast Plus EvaGreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.
Fast Plus Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.
Documents
Specifikationer
Specifikationer
- Förp.:1 mL
- Beskrivning:Fast Plus EvaGreen® qPCR master mix with low Rox, 1 ml
- No. of reactions:100
- Cat. No.:31014-T
- Miljöcertificering:Classified as non-hazardous to aquatic life under CCR title 22 regulations
- Applikation:qPCR dye
- Environmentally Preferable:Safer Human and Environmental Health



