Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, adapter ring, GL 38/430, 1 port for 1/16" tubing, 1 port for 1/8" and 1/16" OD tubing, PP
Katalog # JOURJR-S-13001
Leverantör: VICI JOUR RESEARCH
Gå till:
Produktdetaljer och dokument
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifikationer
- Gänga:GL 38/430
- Beskrivning:Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, adapter ring
- Material:PP
- Seq:06
- Slangkoppling:1 port for 1/16" tubing, 1 port for 1/8" and 1/16" OD tubing