Screw Caps
GL 28
Catalog # HECH41326028
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Svart PP.
Very good chemical resistance
- Heat-resistant up to 110 °C
- Only suitable for bottles 215-2814 to 215-2819
Specifications
- Gänga:GL 28
- Seq:3