Overflow volumetric flasks, class B
420 ml
HECH42115066
DURAN® borosilicate glass 3.3.
- Acid-proof and alkali-resistant markings
- Very good temperature resistance
- Very good chemical resistance
For water analysis.
- Kapacitet:420 ml
- Seq:10
DURAN® borosilicate glass 3.3.
For water analysis.