Orders LinkContactUsLinkComponent
Cellulose, Reference Standards
Cellulose, Reference Standards
Silicified Microcrystalline Cellulose, USP reference standard, Vial, 1 g
Katalog # 1098399.
Leverantör:   USP
Cellulose, Reference Standards
Katalog # 1098399.
Leverantör:  USP
CAS-nummer:  
Cellulose, Reference Standards
Silicified Microcrystalline Cellulose, USP reference standard, Vial, 1 g
CAS-nummer:  
Pris
5 400,00 kr
Log in för tillgänglighet
1 * 1 g
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Dokument
Specifikationer
Specifikationer
  • Förpackningstyp:
    Vial
  • Förp.:
    1 g
  • Standard:
    USP reference standard
  • Beskrivning:
    Silicified Microcrystalline Cellulose
Specifikationer
Rekommendationer
Rekommendationer anpassas endast baserat på dina shoppingpreferenser om du har gett ditt samtycke genom att aktivera "Förbättra min shoppingupplevelse" på sidan "Personlig information".
Annars får du generiska rekommendationer.