Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, GL 45, 1 port for 1/16" tubing, 2 ports for 1/8" and 1/16" OD tubing, PP
Katalog # JOURJR-S-11002
Leverantör: VICI JOUR RESEARCH
Gå till:
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifikationer
- Gänga:GL 45
- Beskrivning:Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter
- Material:PP
- Seq:02
- Slangkoppling:1 port for 1/16" tubing, 2 ports for 1/8" and 1/16" OD tubing