Sample storage tubes, blank and alphanumeric
1,2 ml, Microtubes in grid-referenced microtube cluster rack, 10 racks of 96 tubes/case, Alphanumeric tube
Katalog # 732-4919
Leverantör: THERMO MATRIX TECHNOLOGIES
Sample storage tubes, blank and alphanumeric
Katalog # 732-4919
Leverantör: THERMO MATRIX TECHNOLOGIES
Gå till:
Product Details & Documents
Abgene blank and alphanumeric storage tubes are manufactured from 100% virgin, medical-grade polypropylene. Storage tubes are leak tested to ensure the integrity and security of samples and with a V-bottom design, our alphanumeric storage tubes facilitate sample recovery.
- Highest quality
- Enhanced rack design
Unlike traditional storage tubes or blocks, our blank and alphanumeric storage tubes are available in our specially-designed, stackable, 96-format, microplate-footprint racks to save precious space in your freezer, refrigerator, compound store or on the bench top. In addition, the rack design includes grid referenced lids for simple identification of tube location.
Specifikationer
- Förp.:10 racks of 96 tubes/case
- Typ:Alphanumeric tube
- Kapacitet:1,2 ml
- Beskrivning:Microtubes in grid-referenced microtube cluster rack
- Seq:04
- Cat. No.:732-4919
- Miljöcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact