Gå till:
Produktdetaljer och dokument
Synonymer: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS-nummer:67-64-1
- Formel:CH₃COCH₃
- MDL-nummer:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Kokande Pt:56,2 °C (1013 hPa)
- Smält Pt:–95,4 °C
- Densitet:0,792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Förvaringstemperatur:Omgivande
- EINECS:200-662-2
- FN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
sds_ABSO1007
SDS
Specifikationer
Specifikationer
- Förp.:5 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



