LC-MS nitrogen generators
Low Flow NitroFlow Lab, with air compressor, 10 - 30 l/min, 900×310×700 mm
Katalog # PERCN9306214
Leverantör: PerkinElmer
Gå till:
Produktdetaljer och dokument
Nitrogen generators are designed to supply single or multiple LC-MS instruments with dry nitrogen at purities of 98 – 99,5%. The generator can also be used for solvent evaporation as well as supplying dry nitrogen to analytical instruments.
Specifikationer
- Flödeshastighet:10 - 30 l/min
- B×D×H:900×310×700 mm
- Typ:Low Flow NitroFlow Lab, with air compressor