Orders LinkContactUsLinkComponent
Blanking nuts
Blanking nuts
fit port size 1" NPT
Katalog # COWIPC702.100
Leverantör:   COWIE
Blanking nuts
Katalog # COWIPC702.100
Leverantör:  COWIE
Blanking nuts
Blanking nuts
fit port size 1" NPT
Price
265,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
PTFE, white.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
NPT blanking nuts/plugs are used to seal unused ports. To assemble simply screw the NPT blanking nut into a corresponding NPT port of the reactor lid.
Specifikationer
  • Beskrivning:
    fit port size 1" NPT
  • Seq:
    04