Calibration weight sets, class E2, Proof Line
1 g - 5 kg, Without certificate
Katalog # SARBYCS31-652-AC00
Leverantör: Sartorius Balances
Calibration weight sets, class E2, Proof Line
Katalog # SARBYCS31-652-AC00
Leverantör: Sartorius Balances
Gå till:
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Specifikationer
- Viktset:1 g - 5 kg
- Seq:05
- Typ:Without certificate