Calibration weight sets, class E2, Proof Line
1 mg - 500 mg, Without certificate
Catalog # SARBYCS011-252AC00
Supplier: Sartorius Balances
Calibration weight sets, class E2, Proof Line
Catalog # SARBYCS011-252AC00
Supplier: Sartorius Balances
Jump to:
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Specifications
- Viktset:1 mg - 500 mg
- Seq:01
- Typ:Without certificate