Tank liners, HyPerforma™
Tank liners, 200 l
Catalog # HYCPSH30646.03
Supplier: THERMO BIOPROCESS DIVISION
Jump to:
Product Details & Documents
Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.
- Gusset design allows for a close fit to the tank
- Constructed of CX3-9 film
- For HyPerforma™ Single-Use Mixer DS 300
Specifications
- Beskrivning:Tank liners, 200 l
- Seq:03