Tank liners, HyPerforma™
Tank liners, with liner Ø 630 mm, 50 l
Katalog # HYCPSH30647.01
Leverantör: THERMO BIOPROCESS DIVISION
Gå till:
Product Details & Documents
Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.
- Gusset design allows for a close fit to the tank
- Constructed of CX3-9 film
- For HyPerforma™ Single-Use Mixer DS 300
Specifikationer
- Beskrivning:Tank liners, with liner Ø 630 mm, 50 l
- Seq:04