Accessories for centrifuges
Adapter for Nalgene/Nunc/Corning tubes (max. 250 ml, max. Ø 61 mm)
Katalog # HETT4430
Leverantör: Hettich
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:Adapter for Nalgene/Nunc/Corning tubes (max. 250 ml, max. Ø 61 mm)
- Seq:44
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



