Accessories for centrifuges
Adapter for 12×15 ml tubes (17×120 mm)
HETT5275
: Hettich
- Beskrivning:Adapter for 12×15 ml tubes (17×120 mm)
- Seq:25
- Cat. No.:HETT5275
- Miljöcertificering:ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



