Orders LinkContactUsLinkComponent
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cable with built-in temperature and conductivity and with a port for a pH or ORP sensor (incl. Cable Management Kit), 10 m
Katalog # YSIA6051030-10
Leverantör:   YSI ENVIRONMENTAL
Cables for Multi-Parameter Meter Pro 1030, Handheld
Katalog # YSIA6051030-10
Leverantör:  YSI ENVIRONMENTAL
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cable with built-in temperature and conductivity and with a port for a pH or ORP sensor (incl. Cable Management Kit), 10 m
Price
12 425,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
Specifikationer
  • Beskrivning:
    Cable with built-in temperature and conductivity and with a port for a pH or ORP sensor (incl. Cable Management Kit)
  • Längd:
    10 m
  • Seq:
    03