Odlingsrör
5 ml, PP, without cap, 125 per bag, 75 mm, 12 mm
Catalog # 720-0563
Supplier: Simport Scientific
Jump to:
Product Details & Documents
PP eller PS, graderad, engångs, steril.
- White marking field
- Gradation distinguishing 0,1 (4 ml tube) and 0,25 ml (12 ml tube)
- Biological inert
- Durable, reduces the risk in handling potentially hazardous material
Passar för de flesta rutinapplikationer.
Resist temperatures from −190 to +120 °C (PP) and up to 70 °C (PS).
Max. RCF: 3000×g (PP); 1400×g (PS).
Delivery information: Levereras med eller utan lock av PE.
Documents
Specifications
- Ø:12 mm
- Förp.:125 per bag
- Kapacitet:5 ml
- Beskrivning:PP, without cap
- Höjd:75 mm
- Seq:01



