Sample tubes
2 ml, -, With external thread, graduated, 12,5×49 mm, Self standing
Katalog # SIMPT501-2ATPR
Leverantör: Simport Scientific
Gå till:
Product Details & Documents
PP, non toxic, translucent.
- Very good chemical resistance
- Smooth and uniform inner surface
- Tubes can be autoclaved (121° C) in upright position with caps loosened
- Round bottom tubes only can be centrifuged up to 17000×g
Designed for the storage and transportation of biological material.
*For size only, cap is included.
Certificates
Sök efter LOT-nummerSpecifikationer
- Stängningsstorlek:-
- Typ:Self standing
- Kapacitet:2 ml
- Beskrivning:With external thread, graduated
- Seq:11
- Storlek:12,5×49 mm