Halogen bulbs
Halogen bulb with reflector, 12 V, 50 W
Katalog # ZEIS0000000488372
Leverantör: ZEISS
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:Halogen bulb with reflector
- Seq:905
- Effekt (W):50 W
- Spänning:12 V