Trefot
120×260 mm
Katalog # 451-0161
Leverantör: USBECK Laborgeräte
Gå till:
Produktdetaljer och dokument
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
Specifikationer
- Innerdiam.×H:120×260 mm
- Seq:04