Microtubes
strips of 8 tubes, 0,65 ml
Katalog # RATI8200022
Leverantör: RATIOLAB
Gå till:
Product Details & Documents
Translucent polypropylene (PP) with very low protein binding.
- Very good temperature resistance
- Very good chemical resistance
The 0.65 and 1.2 ml micro-tubes can be supplied as single, strips of 8 or in 96-well format as a refill system for 8 x 12 plates.
Specifikationer
- Kapacitet:0,65 ml
- Beskrivning:strips of 8 tubes