Microtubes
strips of 8 tubes, 1,2 ml
Catalog # RATI8200002
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Translucent polypropylene (PP) with very low protein binding.
- Very good temperature resistance
- Very good chemical resistance
The 0.65 and 1.2 ml micro-tubes can be supplied as single, strips of 8 or in 96-well format as a refill system for 8 x 12 plates.
Specifications
- Kapacitet:1,2 ml
- Beskrivning:strips of 8 tubes