Pycnometer
10 ml
Catalog # HECH42570016
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in stopper
- Good chemical resistance
- Good temperature resistance
Certifications: DIN ISO 3507
Specifications
- Volym (nominell):10 ml